Accredo Safety Gate Valve Lockout, ZING Enterprises
Supplier: ZING Enterprises
7246 7247 7248
76026-330EA 77.75 CAD
76026-330 76026-332 76026-334
Accredo Safety Gate Valve Lockout, ZING Enterprises
Security Locks
Large gate valve lockouts available in 3 sizes.
  • High quality
  • Effective lockout device
  • Very durable
Gate Valve Lockout fits handle diameters from 5'' to 6.5''
Options also include Gate Valve Lockouts that fit handle diameters from 6.5'' to 10'' or 10'' to 13''.
The lockout will accommodate up to 3 padlocks. Includes the legend "DANGER Do Not Remove." The lockout is made from durable plastic and will withstand temperatures from -20 to 200 F.
Certifications: OSHA 1910.147
Order Now
SPECIFICATIONS
Environmentally Preferable