Accredo Safety Group Lock Box, ZING Enterprises
Supplier: ZING Enterprises
7286 7288
76026-386EA 169.8 CAD
76026-386 76026-390
Accredo Safety Group Lock Box, ZING Enterprises
Security Locks
High quality lockout box.
  • High quality
  • Group lockout purposes
  • Very durable
Group Lockout Box, Steel, protects workers in group maintenance and repair operations
Wall-Mountable Group Lockout Box protects workers during maintenance and repair operations. Accommodates up to 12 padlocks, plus 1 on the latch.
The box accommodates up to 7 padlocks.
Certifications: OSHA 1910.147