Accredo Safety Clamp-On Breaker Lockout-cleat, ZING Enterprises
Supplier: ZING Enterprises
7256 7257 7258
76026-350EA 36.26 CAD
76026-350 76026-352 76026-354
Accredo Safety Clamp-On Breaker Lockout-cleat, ZING Enterprises
Security Locks
Convenient way to lockout circuit breakers.
  • No tools required for installation
  • Effective lockout device
  • High quality
120/277V Clamp-On Circuit Breaker Lockout fits circuit breakers 9/16'' W x 3/8'' L
480/600V Clamp-On Circuit Breaker Lockout fits circuit breakers 1-1/16 W x 3/8 L. The lockout measures 1-3/4"H, 2-3/4''L, 1-13/16"W and accommodates 1 padlock, with shackles up to 9/32. The thumbscrew feature allows the device to be installed quickly and easily. The lockout is made from durable plastic (polypropylene) and nylon, and includes a detachable cleat.
480/600V Oversize Clamp-On Circuit Breaker Lockout fits circuit breakers 2-1/4" W x 7/8" L. The lockout measures 2-3/4"H, 2-5/8''L, 3-3/4"W and accommodates 1 padlock, with shackles up to 9/32. The thumbscrew feature allows the device to be installed quickly and easily. The lockout is made from durable plastic (polypropylene) and nylon, and supports OSHA standard 190.147.
The lockout measures 1-3/8"H, 2''L, 7/8"W and accommodates 1 padlock, with shackles up to 9/32. The thumbscrew feature allows the device to be installed quickly and easily. The lockout is made from durable plastic (polypropylene) and nylon.
Certifications: OSHA 1910.147