Accredo Safety Slider Pin Circuit Breaker Lockout, ZING Enterprises
Supplier: ZING Enterprises
7259 7260 7261 7262
76026-356EA 29.63 CAD
76026-356 76026-358 76026-360 76026-362
Accredo Safety Slider Pin Circuit Breaker Lockout, ZING Enterprises
Security Locks
A variety of sizes and configurations are available.
  • No Tools Required for Installation
  • Effective lockout device
  • High quality
120/277 Miniature Circuit Breaker Lockout - Pin In, Standard, provides an effective lockout solution
120/277 Miniature Circuit Breaker Lockout - Pin Out, Standard, provides an effective lockout solution. The lockout measures 3/4"H, 2-3/4''L, 3/4"W and accommodates 1 padlock, with a maximum shackle diameter of 9/32''. The lockout is constructed from durable plastic and is designed for use with ISO/DIN miniature breakers. No tools are required for installation.
120/277 Miniature Circuit Breaker Lockout - Pin Out, Wide, provides an effective lockout solution. The lockout measures 3/4"H, 2-3/4''L, 3/4"W and accommodates 1 padlock, with a maximum shackle diameter of 9/32''. The lockout is constructed from durable plastic and is designed for use with ISO/DIN miniature breakers. No tools are required for installation.
Miniature Circuit Breaker Lockout Set includes 3 miniature circuit breaker lockouts: Pin In Standard, Pin Out Standard, and Pin Out Wide. The lockouts measure 3/4"H, 2-3/4''L, 3/4"W and accommodate 1 padlock each, with a maximum shackle diameter of 9/32''. The lockouts are constructed from durable plastic and are designed for use with ISO/DIN miniature breakers. No tools are required for installation.
Certifications: OSHA 1910.147