ZING Green Safety RecycLockout Group Lockout Box, ZING Enterprises
Supplier: ZING Enterprises
6061R 6061Y
76025-968EA 301.43 CAD
76025-968 76025-970
ZING Green Safety RecycLockout Group Lockout Box, ZING Enterprises
Security Locks
Lockout Box available in red or yellow.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Lockout Box, portable with retractable carry handle, high visible red, accepts 12 padlocks on the box and 1 on the latch, inner shelf provides extra security of stored keys, heavy-duty recycled 18 guage stainless steel, UL Validated, supports sustainability initiatives and LEED credits, measures 6.25"H x 9"W x 4"D, Made in the USA.
Certifications: 29 CFR 1910.147