ZING Green Safety RecycLockout Cable/Hasp Lockout Device, ZING Enterprises
Supplier: ZING Enterprises
7132 7133
76026-272EA 89.51 CAD
76026-272 76026-274
ZING Green Safety RecycLockout Cable/Hasp Lockout Device, ZING Enterprises
Security Locks
Cable/Hasp Lockout is available with 6 ft. or 10 ft. cable.
  • Made in USA
  • Eco-friendly
  • High quality
Cable/Hasp Lockout is designed to secure switches, valves and other energy devices from use while equipment is being serviced or repaired.
Lockout features a 1" inside diameter jaw with pry resistant tabs for extra security. Hasp is high visible red with label "DO NOT OPERATE" and designed with 6 holes to accommodate padlocks with shackles up to 3/8" diameter. Supporting lockout cable is 6 ft or 10 ft. and made of durable steel with coating. Hasp Lockout is constructed from heavy-duty 99% recycled polycarbonate and supports sustainability initiatives and LEED credits. Hasp measures 7"L x 3"W. Made in the USA.
Certifications: 29 CFR 1910.147