ZING Green Safety RecycLockout Gate Valve Lockout, ZING Enterprises
Supplier: ZING Enterprises
7103 7104
76026-222EA 50.02 CAD
76026-222 76026-224
ZING Green Safety RecycLockout Gate Valve Lockout, ZING Enterprises
Security Locks
Designed to cover and secure valve handles from being operated and releasing dangerous pressure.
  • Made in USA
  • 100% recycled plastic
  • High quality
Lockout is high visible red and available in 2 sizes -
Fits 1"-2.5" valves and is designed to accommodate 3 padlocks with shackles up to 3/8" diameter. Lockout is made from heavy-duty 100% recycled polycarbonate that is UL Validated and supports sustainability initiatives and LEED credits. Lockout measures 1"H x 3" diameter. Made in the USA.
Fits 2.5" - 5" valves and is designed to accommodate 3 padlocksMore Product Information