ZING Green Safety Eco Handicap Accessible Right Arrow Label/Decal
Supplier: ZING Enterprises
2716 2717
76024-844PK 26.35 CAD
76024-844 76024-846
ZING Green Safety Eco Handicap Accessible Right Arrow Label/Decal
Signs
The sign measures 6" x 6", and is available in two eco-friendly formats; adhesive labels and window decals.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Adhesive Labels are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
Window Decals (face-adhesive signs) are designed to be placed on the inside of windows. Decals are made from recycled polystyrene.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. Labels and decals are sold 2/pack.
Certifications: ADA Compliance