ZING Green Safety Eco Safety Projecting Sign, Safety Shower
Supplier: ZING Enterprises
2528 2529 2641
76024-042EA 68.97 CAD
76024-042 76024-044 76024-732
ZING Green Safety Eco Safety Projecting Sign, Safety Shower
Signs
Projecting sign is available as a V style, L style, or Corner style.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Projecting V Sign clearly communicates critical safety information
Projecting L Sign clearly communicates critical safety information. The two-sided projecting design significantly increases visibility, and is ideal for designating critical workplace locations. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 7"H x 2.5"W x 7" projecting. Two mounting holes and adhesive tabs are included for easy installation.
Projecting Corner Sign clearly communicates critical safety information. The two-sided projecting design saves space, significantly increases visibility, and is ideal for designating important workplace locations. Can be easily installed on any 90° angle corner. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 10"H x 10"W. Two mounting holes and adhesive tabs are included for easy installation. Recycled materials support sustainability initiatives and LEED credits.
The two-sided projecting design significantly increases visibility, and is ideal for designating important workplace locations. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 7"H x 12"W x 5" projecting. Two mounting holes and adhesive tabs are included for easy installation.