ZING Green Safety Eco Safety Projecting Sign, First Aid
Supplier: ZING Enterprises
2638 2520 2521
76024-726EA 68.18 CAD
76024-726 76025-340 76025-342
ZING Green Safety Eco Safety Projecting Sign, First Aid
Signs
Projecting sign is available as a V style, L style, or Corner style.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Projecting V Sign clearly communicates critical safety information
Projecting L Sign clearly communicates critical safety information. The two-sided projecting design significantly increases visibility, and is ideal for designating critical workplace locations. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 7"H x 2.5"W x 7" projecting. Two mounting holes and adhesive tabs are included for easy installation.
Projecting Corner Sign clearly communicates critical safety information. The two-sided projecting design saves space, significantly increases visibility, and is ideal for designating important workplace locations. Can be easily installed on any 90° angle corner. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 10"H x 10"W. Two mounting holes and adhesive tabs are included for easy installation. Recycled materials support sustainability initiatives and LEED credits.
The two-sided projecting design significantly increases visibility, and is ideal for designating important workplace locations. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 7"H x 12"W x 5" projecting. Two mounting holes and adhesive tabs are included for easy installation.