ZING Green Safety Eco Safety Tri View Sign, Fire Extinguisher
Supplier: ZING Enterprises
3052 3052G 4052 4052G
76025-836EA 96.93 CAD
76025-836 76025-838 76025-908 76025-910
ZING Green Safety Eco Safety Tri View Sign, Fire Extinguisher
Signs
Available in Standard TriView, Slim TriView, Glow Standard TriView, and Glow Slim TriView.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Standard 3-Sided TriView Sign communicates critical safety messages with 180-degree view that significantly increases visibility of information
Standard Glow in the Dark 3-Sided TriView communicates critical safety messages with 180-degree view that significantly increases visibility of information. Glow in the dark feature exceeds 10 hours to meet ASTME 2072-00 standards. Sign is made from high quality 70% post-consumer recycled plastic. Sign measures 7.5"H x 20"W, Projects 5 Inches and includes 2 mounting holes and adhesive tabs. Made in the USA.
ZING Slim 3-Sided TriView Sign communicates critical safety messages with 180-degree view that significantly increases visibility of information. Sign is made from high quality 70% post-consumer recycled plastic. Sign measures 12"H x 9"W, Projects 3 Inches and includes 2 mounting holes and adhesive tabs. Made in the USA.
ZING Slim Glow in the Dark 3-Sided TriView Sign communicates critical safety messages with 180-degree view that significantly increases visibility of information. Glow in the dark feature exceeds 10 hours to meet ASTME 2072-00 standards. Sign is made from high quality 70% post-consumer recycled plastic. Sign measures 12"H x 9"W, Projects 3 Inches and includes 2 mounting holes and adhesive tabs. Made in the USA.
Certifications: ASTME 2072-00