ZING Green Safety Eco Safety Sign, DANGER, Respirable Crystalline Silica
Supplier: ZING Enterprises
1960S 1961 1961A 1961S 2961 2961A 2961S
76023-370EA 26.35 CAD
76023-370 76023-372 76023-374 76023-376 76025-804 76025-806 76025-808
ZING Green Safety Eco Safety Sign, DANGER, Respirable Crystalline Silica
Signs
The sign is available in 2 sizes, 7" x 10", and 10" x 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.