ZING Green Safety Eco Safety Sign, Warning BioHazard Authorized Personnel Only
Supplier: ZING Enterprises
1920S 1921 1921A 1921S
76023-286PK 26.69 CAD
76023-286 76023-288 76023-290 76023-292
ZING Green Safety Eco Safety Sign, Warning BioHazard Authorized Personnel Only
Signs
he sign is available in two sizes, 5" x 7" (2/pack) and 7" x 10", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Self-Adhesive signs and labels are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits.
Certifications: 29 CFR 1910.1030(g)(1)