ZING Green Safety Eco Safety Sign, CAUTION Watch Your Step
Supplier: ZING Enterprises
1154 1154A 1154S 2154 2154A 2154S
76024-890EA 39.94 CAD
76024-890 76024-892 76024-894 76025-232 76025-234 76025-236
ZING Green Safety Eco Safety Sign, CAUTION Watch Your Step
Signs
The sign is available in 2 sizes, 7" x 10", and 10" x 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.
Certifications: OSHA 29 CFR 1910.145