ZING Green Safety Eco Safety Sign, DANGER Gasoline
Supplier: ZING Enterprises
2101 2101A 2101S 1101A 1101S 1101
76023-534EA 47.75 CAD
76023-534 76023-536 76023-538 76024-616 76024-618 76024-974
ZING Green Safety Eco Safety Sign, DANGER Gasoline
Signs
The sign is available in 2 sizes, 7" x 10", and 10" x 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.
Certifications: OSHA 29 CFR 1910.145