ZING Green Safety Eco Safety Sign, Keep Fire Door Closed
Supplier: ZING Enterprises
2083 2083A 2083S 1083 1083A 1083S
76025-092EA 47.75 CAD
76025-092 76025-094 76025-096 76026-446 76026-448 76026-450
ZING Green Safety Eco Safety Sign, Keep Fire Door Closed
Signs
The sign is available in two sizes, 7" x 10" and 10" x 14", and three eco-friendly materials.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.