ZING Green Safety Eco Safety Sign, Not An Exit
Supplier: ZING Enterprises
1080 2080 2080A 2080G 2080S 1080S 1080A 1080G
76022-686EA 39.94 CAD
76022-686 76023-454 76023-456 76023-458 76023-460 76024-492 76026-436 76026-438
ZING Green Safety Eco Safety Sign, Not An Exit
Signs
The sign is available in two sizes, 7" x 10" and 10" x 14", and four eco-friendly materials.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
Glow-in-the-Dark signs are manufactured from recycled plastic with a high-performance, safety grade photoluminescent coating. Signs will glow for a period of 10 hours on a full charge and require no electrical power source.
The signs are pre-drilled for easy mounting, and include adhesive mounting tabs. Impact-resistant plastic is suitable for indoor or outdoor use.
Certifications: OSHA 1910.37(b)(5)