EZBio® Single-Use Media Bottle, PETG, Closed Cap, Sterile
Supplier: Foxx Life Sciences
137-2211-FLS 137-5211-FLS 137-6511-FLS
76018-410PK 745.51 CAD
76018-410 76018-412 76018-414 76018-416 76018-418
EZBio® Single-Use Media Bottle, PETG, Closed Cap, Sterile
Bottles Plastic Bottles
EZBio® single-use PETG bottle assemblies are an innovative and cost efficient option for single-use applications.
  • Perfect for fluid transfer and storage
  • VersaCap® technology
  • More cost efficient than stainless steel apparatuses
  • Gamma irradiated in the United States
  • BPA-free
The EZbio® single-use PETG media bottles are perfect for fluid transfer and storage. Assemblies are made entirely with USP Class VI components. Assemblies take advantage of Foxx’s VersaCap® technology, which means there is no machining necessary to produce the product. Perfect for single-use applications and to phase out costly stainless steel apparatuses that are expensive to clean and validate.
Certifications: All materials comply with USP Class VI standards.