Ezclear® Erlenmeyer Shaker Flasks, Foxx Life Sciences
Supplier: Foxx Life Sciences
248-4131-OEM 248-4132-OEM 248-4142-OEM 248-4231-OEM 248-4232-OEM 248-4242-OEM
76018-112EA 120.22 CAD
76018-112 76018-114 76018-116 76018-118 76018-120 76018-122
Ezclear® Erlenmeyer Shaker Flasks, Foxx Life Sciences
Flasks Erlenmeyer Flasks
EZclear® Erlenmeyer Shaker Flasks are molded from virgin, optically clear, non-leaching polycarbonate resin. These flasks retain clarity after gamma sterilization, and comply with stringent FDA and USP requirements.
  • FDA and USP compatible
  • Fits all standard clamps used by a standard shaking incubator
  • Autoclavable
  • Pyrogen-free
  • Dnase/Rnase-free
  • Available with standard or vented VersaCap® closure
Sterile EZclear® Erlenmeyer Shaker Flasks are certified as non-pyrogenic and DNase/RNase-free, making them ideal for cell cultures, microbial cultures, plants, media preparation, storage, and all other applications
EZclear® Flasks are available with a flat or baffled base for increased oxygenation and mixing. Flasks are compatible with all 53mm VersaCap® technology and adapter inserts, enabling easy customization of top connections. Vented VersaCap® with a 0.2 µm hydrophobic membrane can be purchased with the unit, or separately as an accessory. All flasks and VersaCaps® are autoclavable at 15 psi and 121 °C for 20 minutes.
Flasks fit all standard clamps. They are sterility tested to SAL 10-6, and made from Class VI autoclavable materials. Manufactured in a class 100,000 cleanroom.
They also fit all standard shaking incubator clamps and can be placed on any platform.