SP Wilmad-LabGlass Gas Washing Bottles with Fritted Discs and [ST] 40/35 Stopper Assembly, SP Industries
Supplier: SP Industries
LG-3765-126 LG-3765-130 LG-3765-124
14207-580EA 1131.17 CAD
14207-580 14209-050 14209-668
SP Wilmad-LabGlass Gas Washing Bottles with Fritted Discs and [ST] 40/35 Stopper Assembly, SP Industries
Bottles Gas Washing Bottles
These borosilicate glass gas washing bottles are generally used to saturate or dry gas streams.
Complete units consist of a bottle and a standard taper 40/35 stopper assembly with an inlet tube that is sealed through the center of a coarse (C) fritted glass disc.
Inlet tube extends nearly to the base of the bottle.