LB Broth Miller Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
LB Broth Miller is a granulated medium for the cultivation of E. coli.
- Granulation preserves homogeneity
- Easy to weigh out
- Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.
E. coli on scales ranging from small cultures to fermentation.