ZING Green Safety Eco Safety Sign, Exit, White on Red
Supplier: ZING Enterprises
1078 1078A 1078G 1078S 2078 2078A 2078G 2078S
76024-428EA 39.94 CAD
76024-428 76024-430 76024-432 76024-434 76024-452 76024-454 76024-456 76024-458
ZING Green Safety Eco Safety Sign, Exit, White on Red
Signs
The sign is available in two sizes, 7" x 10" and 10" x 14", and four eco-friendly materials.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
Glow-in-the-Dark signs are manufactured from recycled plastic with a high-performance, safety grade photoluminescent coating. Signs will glow for a period of 10 hours on a full charge and require no electrical power source.
The signs are pre-drilled for easy mounting, and include adhesive mounting tabs. Impact-resistant plastic is suitable for indoor or outdoor use.
Certifications: OSHA 1910.37,ASTM E2072 - 00