SHOWA 8814 Heat Resistant Glove, Showa
Supplier: Showa
8814-07 8814-08 8814-10
89022-343CS 453.35 CAD
89022-343 89022-345 89022-349
SHOWA 8814 Heat Resistant Glove, Showa
Gloves
The SHOWA 8814 heat-resistant gloves are designed to withstand temperatures up to 500°F/260°C. Even after multiple washings, the 8814 maintains its heat-protective qualities.
  • The textured surface of the neoprene coating provides superior tactile dexterity
  • Easy donning and doffing makes the glove easy to remove in an emergency
  • Machine washable
  • Rough grip, spray coated neoprene
Gloves are used in high heat applications. Use when handling castings, glassware, or bakeware.