SHOWA 874 Butyl II, Chemical Resistant Gloves, Showa
Supplier: Showa
874R-07 874-07 874R-08 874R-07 874R-09 874R-10 874R-11 874R-09 874R-08
28888-320PK 308.4 CAD
28888-320 28888-338 28888-402 CA28888-320 CA28888-322 CA28888-324 CA28888-326 28888-322 CA28888-402
SHOWA 874 Butyl II, Chemical Resistant Gloves, Showa
Gloves
Butyl II rubber gloves provide superior resistance to highly corrosive acids, amines, ketones, and esters
  • Butyl 14 mil
  • Unlined butyl rubber
  • Hazmat glove
The SHOWA 874 Butyl II gloves offer the highest permeation resistance to gases and water vapors of any protective material used to make gloves. They are ideally suited for use with highly toxic chemicals. Used with Hazmat A suits. Gloves are available in smooth or rough finishes. Smooth finish gloves feature a gauntlet cuff style, and rough finish gloves have a rolled cuff.
They are ideally suited for use in Hazmat situations.
Certifications: TAA compliant product.