SHOWA 8110 A5 Cut Resistant , Uncoated, HPPE Glove, Showa
Supplier: Showa
8110-07 8110-08 8110-09 8110-10
47742-304CS 642.1 CAD
47742-304 47742-306 47742-308 47742-310 47742-312
SHOWA 8110 A5 Cut Resistant , Uncoated, HPPE Glove, Showa
Gloves
A5 Cut resistant rating is perfect for hazardous cut applications.
  • Wire-free
  • Made in U.S.
These gloves offer extreme protection from cuts. Launderable for longer wear.
Certifications: ANSI Cut Level A5.