SHOWA 724R PVC, Chemical Resistant, 14" Gauntlet Glove, Showa
Supplier: Showa
724R-10
47742-280CS 1590 CAD
47742-280
SHOWA 724R PVC, Chemical Resistant, 14" Gauntlet Glove, Showa
Gloves
Tough PVC coating is designed for a wide variety of chemical, maintenance, and construction applications.
  • PVC, fully coated
  • Cotton Interlocked knit lined
  • Rough grip
These gloves are designed for tough, abrasive applications, and for handling chemicals.
Certifications: ANSI Abrasion Level 4.