Avantor® Prevail C18-Select, HPLC Columns
Supplier: Avantor
99301 99316 99300 99119/N
75797-784EA 2358.64 CAD
75797-784 75798-954 75798-956 75798-960 75830-688 75830-692
Avantor® Prevail C18-Select, HPLC Columns
Chromatography Columns
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases.
  • Stable from highly organic to highly aqueous
  • Specialty phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
  • USP L1
Same benefits as Avantor® Hichrom Prevail™ C18, but slightly more retentive.
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis ofMore Product Information