Avantor® Prevail C18, HPLC Columns
Supplier: Avantor
43818 99200 99322 43829 99202 99204 99279 43903 99208 99209 99210 99211 96683/N 99286/N
75797-758EA 1445.87 CAD
75797-758 75797-760 75797-762 75797-764 75797-766 75797-768 75797-770 75797-772 75797-774 75797-776 75797-780 75798-914 75798-916 75798-920 75798-922 75798-924 75798-926 75798-928 75798-930 75812-872 75812-874 75813-100 75830-682 75830-684 75830-704 76402-316 76402-320 76402-322 76402-324
Avantor® Prevail C18, HPLC Columns
Chromatography Columns
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases.
  • Stable from highly organic to highly aqueous
  • Specialty phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
  • USP L1
Flexibility to switch between varied mobile phase conditions to suit a variety of applications. Excellent sensitivity for microbore applications.
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
USP code L1 L1 L1 L1