Avantor® Prevail Amide, HPLC Columns
Supplier: Avantor
88660 88665
75797-738EA 2529.77 CAD
75797-738 75797-740 76402-318
Avantor® Prevail Amide, HPLC Columns
Chromatography Columns
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases.
  • Stable from highly organic to highly aqueous
  • Specialty phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
Polar embedded phase.
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
Order Now
SPECIFICATIONS
Environmentally Preferable