SP Wilmad-LabGlass Freeze Drying Flasks, SP Industries
Supplier: SP Industries
LG-13100-108 LG-13100-100 LG-13100-106 LG-13100-112 LG-13100-104 LG-13100-110 LG-13100-102
14007-770EA 230.53 CAD
14007-770 14007-768 14007-766 14007-764 14007-762 14007-774 14007-772
SP Wilmad-LabGlass Freeze Drying Flasks, SP Industries
Flasks Flat Bottom Flasks
These borosilicate glass freeze drying replacement flasks for VirTis® lyophilizing equipment have extra-wide openings for efficient shell freezing.
  • Suited for bulky samples
Wall thickness of flask is designed for vacuum applications.