TeacherGeek Small Motor with Leads and Mount
Supplier: TEACHERGEEK INC
1821-01 1821-02
470229-098EA 4.75 CAD
470229-098 470229-100
TeacherGeek Small Motor with Leads and Mount
Educational Materials Physics Educational Materials TeacherGeek System Components
Do you have a contraption to power? Here is a simple solution.
  • The motor turns at the perfect speed for most projects
  • Use these for motor powered, cars, boats, scribble bots, and other inventions
This 1.5-3V motor with alligator clip leads that already has a motor mount and adapter pin attached. Also included is a AA single battery holder. Squeeze the alligator clips to attach them to the battery holder terminals. Turn the motor on and off by attaching and detaching the leads. Motor Shaft Diameter: 2.3mm.
Please note: You may need to give the motorMore Product Information