Vydac 201TP C18, HPLC Columns
Supplier: Avantor
201TP5410 201TP5215 201TP5315 201TP5415 201TP54 201TP104 201TP5203 201GD104/N 201GD52/N 201GK104/N 201GK52/N 201GK54/N
75798-250EA 2938.34 CAD
75798-250 75798-252 75798-254 75798-256 75798-258 75798-260 75798-262 75798-264 75798-266 75798-268 75798-270 75798-274 75798-282 75798-284 75830-722 76176-938 76398-012 76398-014 76398-016 76398-018 76398-020 76398-022 76398-024 76398-026
Vydac 201TP C18, HPLC Columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L1
Non-endcapped C18 phase. Developed for the separation of PAHs.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registeredMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Carbon load 8%
Endcapped No
USP code L1