Vydac 238TP C18, HPLC Columns
Supplier: HICHROM LIMITED
238TP53 238TP54 238TP510 238GK52/N 238GK54/N
75798-528EA 2186.55 CAD
75798-528 75798-530 75798-532 75798-534 75798-536 75798-550 75798-552 75798-554 75798-556 75798-558 75798-560 75798-562 75798-564 75798-566 75798-568 75798-570 75798-572 75798-574 75798-776 75813-056 75830-672 75830-674 75830-680 75831-442 75831-444 75831-452 76398-140 76398-146
Vydac 238TP C18, HPLC Columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L1
First generation monomeric C18 phase. Recommended for the same applications as 218TP but offers alternative C18 selectivity.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.