Vydac Everest C18, HPLC Columns
Supplier: Avantor
238EV5215 238EV5.07515 238EV5C0110 238EV5.1515 238EV5.1525 238EV1022 238GD54EV/N 238GK54EV/N 238GD52EV/N
75797-968EA 1459.29 CAD
75797-968 75797-970 75797-972 75798-688 75798-690 75798-692 75798-694 75798-698 75798-700 75798-702 75798-704 75799-030 75799-032 75799-034 75799-036 75799-038 75799-040 75799-042 75799-044 75799-046 75799-048 75799-050 75799-052 75799-054 75799-056 75812-964 75812-966 75812-968 75830-396 75830-668 75830-788 75831-448 76398-138
Vydac Everest C18, HPLC Columns
Chromatography Columns
Everest columns have unique selectivity and sensitivity, which are the result of bonding technology that improves C18 surface coverage and deactivates residual silanols.
  • Unique selectivity for hydrophilic and hydrophobic peptides
  • High resolution for complex peptide digests
  • Excellent sensitivity with little or no TFA in mobile phase
  • USP L1
Vydac 238EV (C18) offers maximum surface coverage for the highest resolution of complex samples. It is recommended for the analysis of complex enzymatic digests (>12 proteins).
Previously, the best 300 Å C18More Product Information