BD BBL™ Sabouraud Dextrose Agar, Prepared Media Stacker™ Plates, BD Diagnostics
Supplier: Avantor
CA-90001-284PK
214.64
CAD
CA-90001-284
CA90001-252
CA90008-856
BD BBL™ Sabouraud Dextrose Agar, Prepared Media Stacker™ Plates, BD Diagnostics
Media for Microbiology
Prepared Media for Microbiology
Prepared plated media are designed for the isolation of microorganisms from samples or specimens and come in 100×15 mm petri-style configurations.
- Plates contain a single medium
- Interlocking, stackable design to minimize sliding or sample disruption
- Ready for immediate use
Store at 2 to 8 °C (36 to 46 °F).