2YT Broth Bottled Media, Teknova
Supplier: TEKNOVA CA
Y0167 Y0166
CA100219-100EA 179.61 CAD
CA100219-100 CA100220-314
2YT Broth Bottled Media, Teknova
Media for Microbiology Prepared Media for Microbiology
2YT broth.
  • 500 ml and 1 Liter
  • Sterile
Contains 1.6% tryptone, 1.0% yeast extract, 0.5% NaCl. Ambient shipping conditions. Store at room temperature.