Tryptic Soy Broth, Teknova
Supplier: TEKNOVA CA
T1550
T1515
T1516
T9400
CA100218-958EA
166.69
CAD
CA100218-958
CA100220-230
CA101320-664
CA101320-708
Tryptic Soy Broth, Teknova
Media for Microbiology
Prepared Media for Microbiology
Tryptic Soy Broth is a general purpose media for growth of bacteria that have high nutritional requirements.
- Ambient shipping conditions
- Store at room temperature
Contains 1.7% tryptone, 0.3% soytone, 0.25% Glucose, 0.5% sodium chloride, and 0.25% potassium phosphate.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...