Toledo Vascular Micro Scissors, Sklar
Supplier: Sklar
98-9410 98-9411 98-9412 98-9413 98-9414 98-9415 98-9416 98-9417 98-9418 98-9419 98-9442 98-9443 98-9444 98-9445 98-9446 98-9447 98-9448 98-9449 98-9427 98-9428 98-9429 98-9435 98-9436 98-9437 98-9438 98-9439 98-9425 98-9426 98-9440 98-9441
75995-628EA 1344.03 CAD
75995-628 75995-630 75995-632 75995-634 75995-636 75995-638 75995-640 75995-642 75995-644 75995-646 75995-648 75995-650 75995-652 75995-654 75995-656 75995-658 75995-660 75995-662 75995-752 75995-754 75995-756 75995-758 75995-760 75995-762 75995-764 75995-766 75996-050 75996-052 75996-054 75996-056
Toledo Vascular Micro Scissors, Sklar
Scissors
Sklar's® Toledo Micro Vascular Scissors are most commonly used to repair the smallest vessels in small, difficult to reach surgical sites during vascular procedures.
  • Reusable
  • Nonsterile
The jaws of these scissors are extremely small and angled. Some models have one probe-pointed blade, which is useful if dissection is necessary. The second blade is sharp.