Toledo Micro Forceps, Sklar
Supplier: Sklar
98-9205 98-9206 98-9207 98-9208 98-9211 98-9212 98-9213 98-9214 98-9216 98-9218 98-9221 98-9222 98-9223 98-9224 98-9226 98-9228 98-9201 98-9202 98-9203 98-9204
75995-958EA 2971.24 CAD
75995-958 75995-960 75995-962 75995-964 75995-966 75995-968 75995-970 75995-972 75995-974 75995-976 75995-978 75995-980 75995-982 75995-984 75995-986 75995-988 75996-362 75996-364 75996-366 75996-368
Toledo Micro Forceps, Sklar
Forceps
Sklar's® Toledo Micro Forceps are commonly used for holding and dissection of small, fine vessels and membranes.
  • Reusable
  • Nonsterile
The forceps are available straight, curved, or angled, and with or without teeth. The handle has semi-cylandrical, ergonomicly designed, texturized grips for optimal comfort and utilization. This style of handle allows the user to change direction and angle of the instrument by rolling, which is particularly useful during dissection.