Adventitia Scissors, Sklar
Supplier: Sklar
98-7980 98-7988
75996-338EA 1780.75 CAD
75996-338 75996-342
Adventitia Scissors, Sklar
Scissors
Sklar® Adventitia Scissors are double spring leaf scissors commonly used for trimming tissue that has already been dissected.
  • Reusable
  • Nonsterile
These scissors should only be used for trimming, not dissecting, and are best used under high magnification due to their extremely sharp tips and blades.
Order Now
SPECIFICATIONS
Environmentally Preferable