Hook, Sklar
Supplier: Sklar
98-7321 98-7322 98-7323
75995-558EA 509.83 CAD
75995-558 75995-560 75995-562
Hook, Sklar
Retractors
The Sklar® Hook is designed for use in manipulation of brain tissue during surgery.
  • Reusable
  • Nonsterile
The hook is available with a blunt or semi-sharp tip, angled at 45 or 90 degrees.