Yasargil Raspatory, Sklar
Supplier: Sklar
98-2120 98-2121 98-2122 98-2123 98-2124 98-2125 98-2126 98-2127
75995-856EA 646.75 CAD
75995-856 75995-858 75995-860 75995-862 75995-864 75995-866 75995-868 75995-870
Yasargil Raspatory, Sklar
Raspatories
Yasargil raspatory is a micro neurosurgery file used to scrape or smooth down bone surfaces so as not to cause damage to surrounding areas.
  • Reusable and nonsterile
  • Round handle
  • Variety of tip configurations available
This round handle instrument is available in a variety of tip configurations, and straight, bayonet or S-curve shaft.