Econo™ Non sterile Multi-Cut Utility Scissors, Disposable, Sklar
Supplier: Sklar
96-5390 96-5391 96-5392 96-5393 96-5394 96-5395 96-5396 96-5397 96-5398 96-5399
75996-062CS 816.76 CAD
75996-062 75996-064 75996-066 75996-068 75996-070 75996-072 75996-074 75996-076 75996-078 75996-080
Econo™ Non sterile Multi-Cut Utility Scissors, Disposable, Sklar
Scissors
Econo™ Utility Scissors are a multipurpose instrument used for cutting nonsterile materials such as dressings, casts, and surgical drapes.
  • Flattened, wide, blunt tip on serrated blade greatly reduces likelihood of skin being nicked
  • One larger plastic finger ring for maximized control while cutting
These scissors are angled and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting. Econo™ instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.