Disposable Multi-Cut Utility Scissors, Floor Grade, Sterile, Sklar
Supplier: Sklar
96-1220 96-1222 96-1223 96-1224 96-1225 96-1226 96-1227 96-1228 96-1229
75995-294CS 502.59 CAD
75995-294 75995-296 75995-298 75995-300 75995-302 75995-304 75995-306 75995-308 75995-310
Disposable Multi-Cut Utility Scissors, Floor Grade, Sterile, Sklar
Scissors
Sklar® Utility Scissors are a multipurpose instrument used for cutting nonsterile materials such as dressings, casts, and surgical drapes.
  • Flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked
  • One larger plastic finger ring for maximized control while cutting
These scissors are angled and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient,More Product Information