NZYCM Broth Bottled Media, Sterile, Teknova
Supplier: TEKNOVA CA
N0163
CA100219-262EA 466.48 CAD
CA100219-262
NZYCM Broth Bottled Media, Sterile, Teknova
Media for Microbiology Prepared Media for Microbiology
For the maintenance and propagation of <italic>E
  • 1 Liter
  • Sterile
Contains 1.0% Casein digest (NZ amine), 0.1% casamino acids, 0.5% yeast extract, 0.5% NaCl, 10mM MgSO4. Ambient shipping conditions. Store at room temperature.
coli.</italic>