ReproLife™ Reproductive Medium Complete Kit, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1097 LM-0044 LL-0068
103389-426EA 235.38 CAD
103389-426 103389-428 103389-434
ReproLife™ Reproductive Medium Complete Kit, Lifeline Cell Technology
Cell Culture Media
Optimized for the culture of human female reproductive epithelial cells.
  • Supports the growth of these cells over 5 population doublings
  • No antimicrobials or phenol red
  • Complete Kit to prepare fresh medium
ReproLife™ is optimized for the culture of Normal Human Female Reproductive Epithelial Cells, ReproLife™ supports the growth of these cells over a period of at least 5 population doublings.
ReproLife™ is offered in a kit format composed of basal medium and associated supplements and growth factors called “LifeFactors®.” This kit allows preparation of fresh medium in your laboratory, extending shelf life and enhancing performance. ReproLife™ Basal Medium is provided in a light-protected 500 mL bottle. The remaining LifeFactors® are packaged in a convenient gas-impermeable pouch for easy storage.
The LifeFactors® Kit includes: L-glutamine 15 mL, Epinephrine 0.5 mL, Hydrocortisone hemisuccinate 0.5 mL, rh EGF 1 mL, Extract P™ 4 mL, Apo-Transferrin 1 mL, rh Insulin 1 mL, Gentamicin and Amphotericin B 0.5 mL.