LB Broth with 100 µg/mL Ampicillin Bottled Media, Teknova
Supplier: Teknova
L8105 L8105
CA100219-742EA 364.71 CAD
CA100219-742 100219-742
LB Broth with 100 µg/mL Ampicillin Bottled Media, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with recombinant strains of E.coli containing plasmids with the resistance gene to Ampicillin (bla or ampR) such as pBluescript, pGEM, or the pUC series of plasmids.
  • 1 Liter
  • Sterile
Ingredients: 1.0% Tryptone (10.0g/L), 0.5% Yeast Extract (5.0g/L), 1.0% Sodium chloride (NaCl) (10.0g/L), 100µg/mL Ampicillin. Ship on ice. Store frozen.