LB Broth with 7.5% Glycerol Bottled Media, Sterile, Teknova
Supplier: Teknova
L8075
CA100219-614EA 494.95 CAD
CA100219-614
LB Broth with 7.5% Glycerol Bottled Media, Sterile, Teknova
Media for Microbiology Prepared Media for Microbiology
For the maintenance and propagation of E.coli.
  • Excellent media for plasmid DNA production
  • 1 Liter
  • Sterile
Contains 1.0% Tryptone, 0.5% yeast extract, 1.0% NaCl, 7.5% glycerol. Ambient shipping conditions. Store at room temperature.