BCR® Stainless Steel Flat Mop Sieve, Berkshire
Supplier: Berkshire
BCR®
BCRFMSIEVE1
76027-822CS 546.3 CAD
76027-822
BCR® Stainless Steel Flat Mop Sieve, Berkshire
Mops
The Flat Mop Sieve is made of high quality stainless steel, and is suitable for the one-step wet cleaning process.
  • High quality stainless steel
  • Fully autoclavable
Use to release excess waste water into a containment bucket. Simply press the mop onto the sieve to release excess cleaning solution, and apply to surface. Sieve is fully autoclavable.
Certifications: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.