CM Raffinose Broth minus Uracil Bottled Media, Teknova
Supplier: TEKNOVA CA
C9221 C9221
CA101319-784EA 614.84 CAD
CA101319-784 101319-784
CM Raffinose Broth minus Uracil Bottled Media, Teknova
Media for Microbiology Prepared Media for Microbiology
Carbon: 2% Raffinose, Nitrogen: 0.5% Ammonium Sulfate, Minus: Uracil.
  • 1 Liter
  • Sterile
Ambient shipping conditions. Refrigerate to store.